siRNA Details
| Gene Symbol: SNAPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006049.2 | NP_006040.1 | 3 | 312 | ||
| GenBank mRNA | AF093593.1 | - | 3 | 235 | ||
| AK024763.1 | - | 3 | 711 | |||
| AK292051.1 | - | 2 | 160 | |||
| AK312018.1 | - | 3 | 284 | |||
| BC014315.1 | - | 2 | 180 | 3997264 | ||
| CD685245.1 | - | 3 | 349 | |||
| CN347010.1 | - | 3 | 234 | |||
| DB037177.1 | - | 3 | 244 | |||

SNAPC5
Genomic Map