siRNA Details
| Gene Symbol: TCIRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006019.3 | NP_006010.2 | 14 | 1709 | ||
| NM_006053.3 | NP_006044.1 | 9 | 1468 | |||
| XM_005273709.3 | XP_005273766.1 | 15 | 1868 | |||
| XM_011544726.2 | XP_011543028.1 | 14 | 1608 | |||
| XM_017017089.1 | XP_016872578.1 | 14 | 1704 | |||
| XM_017017090.1 | XP_016872579.1 | 13 | 2047 | |||
| GenBank mRNA | AF025374.1 | - | 9 | 1468 | ||
| AY548968.1 | - | 2 | 330 | |||
| BC018133.1 | - | 14 | 1688 | 3905446 | ||
| BC032465.1 | - | 14 | 1681 | 5210733 | ||
| U45285.1 | - | 14 | 1637 | |||

TCIRG1
Genomic Map