siRNA Details
| Gene Symbol: BTN2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197237.1 | NP_001184166.1 | 8 | 1201 | ||
| NM_001197239.1 | NP_001184168.1 | 6 | 549 | |||
| NM_001197240.1 | NP_001184169.1 | 7 | 1050 | |||
| NM_006995.4 | NP_008926.2 | 8 | 1179 | |||
| NM_181531.2 | NP_853509.1 | 7 | 831 | |||
| XM_005248797.3 | XP_005248854.1 | 8 | 1196 | |||
| XM_005248798.4 | XP_005248855.1 | 7 | 1156 | |||
| XM_006714953.3 | XP_006715016.1 | 8 | 1448 | |||
| XM_006714955.3 | XP_006715018.1 | 8 | 1425 | |||
| XM_011514228.2 | XP_011512530.1 | 8 | 1501 | |||
| XM_011514229.2 | XP_011512531.1 | 8 | 1010 | |||
| XM_011514231.2 | XP_011512533.1 | 6 | 874 | |||
| XM_017010166.1 | XP_016865655.1 | 8 | 1199 | |||
| XM_017010167.1 | XP_016865656.1 | 7 | 1101 | |||
| XM_017010168.1 | XP_016865657.1 | 8 | 1681 | |||
| XM_017010169.1 | XP_016865658.1 | 8 | 1399 | |||
| GenBank mRNA | AK123902.1 | - | 3 | 1791 | ||
| AK289883.1 | - | 7 | 819 | |||
| AK293526.1 | - | 6 | 524 | |||
| BC013258.2 | - | 6 | 751 | |||
| BC014021.2 | - | 8 | 1177 | 4124138 | ||
| BI754184.1 | - | 6 | 525 | |||
| U90550.1 | - | 8 | 1154 | |||

BTN2A2
Genomic Map