siRNA Details
| Gene Symbol: TRIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001248006.1 | NP_001234935.1 | 6 | 1539 | ||
| NM_001248007.1 | NP_001234936.1 | 5 | 1202 | |||
| NM_006458.3 | NP_006449.2 | 7 | 1540 | |||
| NM_033278.3 | NP_150594.2 | 6 | 1375 | |||
| XM_011519844.1 | XP_011518146.1 | 7 | 1639 | |||
| XM_011519845.2 | XP_011518147.1 | 6 | 1320 | |||
| XM_017017098.1 | XP_016872587.1 | 6 | 1351 | |||
| XM_017017099.1 | XP_016872588.1 | 6 | 1243 | |||
| GenBank mRNA | AF045239.1 | - | 5 | 1144 | ||
| AF220020.1 | - | 5 | 1144 | |||
| AF220021.1 | - | 6 | 1111 | |||
| AF220022.1 | - | 5 | 907 | |||
| AK126685.1 | - | 4 | 768 | |||
| AK291396.1 | - | 6 | 1539 | |||
| AK298843.1 | - | 5 | 1078 | |||
| AK299586.1 | - | 6 | 1332 | |||
| AK311127.1 | - | 6 | 1330 | |||
| AL360185.1 | - | 1 | 30 | |||
| AL360192.1 | - | 6 | 1081 | |||
| BC096827.1 | - | 6 | 1375 | 6108991 | ||
| DA347370.1 | - | 1 | 405 | |||

TRIM3
Genomic Map