siRNA Details
| Gene Symbol: RAD51AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130862.1 | NP_001124334.1 | 2, 3 | 216 | ||
| NM_006479.4 | NP_006470.1 | 2, 3 | 216 | |||
| GenBank mRNA | AF006259.1 | - | 2, 3 | 111 | ||
| AK096930.1 | - | 2, 3 | 93 | |||
| AK290428.1 | - | 2, 3 | 110 | |||
| AK291948.1 | - | 2, 3 | 110 | |||
| AK300773.1 | - | 2, 3 | 110 | |||
| AK316367.1 | - | 2, 3 | 117 | |||
| AL832243.1 | - | 2, 3 | 94 | |||
| BC006992.1 | - | 2, 3 | 110 | 3997352 | ||
| BC016330.1 | - | 2, 3 | 108 | 4107592 | ||
| DC397655.1 | - | 2, 3 | 216 | |||

RAD51AP1
Genomic Map