siRNA Details
| Gene Symbol: DLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016941.3 | NP_058637.1 | 2 | 238 | ||
| NM_203486.2 | NP_982353.1 | 2 | 238 | |||
| GenBank mRNA | AK075302.1 | - | 2 | 208 | ||
| AK308460.1 | - | 2 | 260 | |||
| BC000218.2 | - | 2 | 187 | 3508262 | ||
| DB035825.1 | - | 2 | 238 | |||

DLL3
Genomic Map