siRNA Details
| Gene Symbol: PTTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006607.2 | NP_006598.2 | 1 | 188 | ||
| GenBank mRNA | AF095288.1 | - | 1 | 188 | ||
| BC069114.1 | - | 1 | 188 | 7216932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTTG2| Gene Symbol: PTTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006607.2 | NP_006598.2 | 1 | 188 | ||
| GenBank mRNA | AF095288.1 | - | 1 | 188 | ||
| BC069114.1 | - | 1 | 188 | 7216932 | ||