siRNA Details
| Gene Symbol: C1QL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006688.4 | NP_006679.1 | 1 | 801 | ||
| GenBank mRNA | AF095154.1 | - | 1 | 586 | ||
| BC008798.2 | - | 1 | 801 | 3635430 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1QL1| Gene Symbol: C1QL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006688.4 | NP_006679.1 | 1 | 801 | ||
| GenBank mRNA | AF095154.1 | - | 1 | 586 | ||
| BC008798.2 | - | 1 | 801 | 3635430 | ||