siRNA Details
| Gene Symbol: BTNL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197975.2 | NP_932079.1 | 2 | 522 | ||
| GenBank mRNA | AB020625.1 | - | 2 | 496 | ||
| AK025267.1 | - | 2 | 500 | |||
| AY358385.1 | - | 2 | 471 | |||
| BC100800.1 | - | 2 | 408 | 40000697 | ||
| BC100801.1 | - | 2 | 408 | 40000698 | ||
| BC100802.1 | - | 2 | 408 | 40000699 | ||
| DA431389.1 | - | 2 | 522 | |||

BTNL3
Genomic Map