siRNA Details
| Gene Symbol: RAB40B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006822.2 | NP_006813.1 | 1 | 162 | ||
| XM_005256334.4 | XP_005256391.1 | 1 | 142 | |||
| XM_005256335.3 | XP_005256392.1 | 1 | 134 | |||
| XM_017024042.1 | XP_016879531.1 | 1 | 109 | |||
| GenBank mRNA | AB209711.1 | - | 1 | 140 | ||
| AL047483.1 | - | 1 | 140 | |||
| BC018039.1 | - | 1 | 162 | 4801271 | ||
| U05227.1 | - | 1 | 81 | |||

RAB40B
Genomic Map