siRNA Details
| Gene Symbol: ILVBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006844.4 | NP_006835.2 | 16 | 2001 | ||
| XM_005259717.4 | XP_005259774.1 | 16 | 2148 | |||
| XM_011527651.2 | XP_011525953.1 | 15 | 1980 | |||
| GenBank mRNA | AB209065.1 | - | 10 | 3928 | ||
| BC000109.1 | - | 5 | 623 | |||
| BC011722.2 | - | 16 | 1968 | 4300207 | ||
| BC011761.2 | - | 16 | 1894 | 3622453 | ||
| BM676342.1 | - | 1 | 343 | |||
| U61263.1 | - | 16 | 1876 | |||

ILVBL
Genomic Map