siRNA Details
| Gene Symbol: ILVBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006844.4 | NP_006835.2 | 3 | 354 | ||
| XM_005259717.4 | XP_005259774.1 | 3 | 501 | |||
| XM_011527651.2 | XP_011525953.1 | 2 | 333 | |||
| GenBank mRNA | AB209065.1 | - | 1 | 523 | ||
| BC000109.1 | - | 2 | 298 | |||
| BC011722.2 | - | 3 | 321 | 4300207 | ||
| BC011761.2 | - | 3 | 247 | 3622453 | ||
| BC126913.1 | - | 1 | 94 | |||
| HY011076.1 | - | 3 | 354 | |||
| U61263.1 | - | 3 | 229 | |||

ILVBL
Genomic Map