siRNA Details
| Gene Symbol: CCDC85B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006848.2 | NP_006839.2 | 1 | 521 | ||
| GenBank mRNA | AK312109.1 | - | 1 | 281 | ||
| BC008796.2 | - | 1 | 286 | 3617697 | ||
| U63825.1 | - | 1 | 169 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC85B| Gene Symbol: CCDC85B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006848.2 | NP_006839.2 | 1 | 521 | ||
| GenBank mRNA | AK312109.1 | - | 1 | 281 | ||
| BC008796.2 | - | 1 | 286 | 3617697 | ||
| U63825.1 | - | 1 | 169 | |||