siRNA Details
Gene Symbol: ADRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281437.1 | NP_001268366.1 | 7 | 910 | ||
NM_001281438.1 | NP_001268367.1 | 7 | 914 | |||
NM_007002.3 | NP_008933.2 | 8 | 1031 | |||
NM_175573.2 | NP_783163.1 | 8 | 1027 | |||
XM_005260257.1 | XP_005260314.1 | 9 | 1118 | |||
XM_011528503.1 | XP_011526805.1 | 8 | 1866 | |||
XM_017027602.1 | XP_016883091.1 | 8 | 1887 | |||
XM_017027603.1 | XP_016883092.1 | 9 | 1139 | |||
XM_017027604.1 | XP_016883093.1 | 8 | 1883 | |||
XM_017027605.1 | XP_016883094.1 | 8 | 1887 | |||
XM_017027606.1 | XP_016883095.1 | 7 | 1770 | |||
XM_017027607.1 | XP_016883096.1 | 7 | 1766 | |||
XM_017027608.1 | XP_016883097.1 | 6 | 1556 | |||
XM_017027609.1 | XP_016883098.1 | 6 | 1535 | |||
XM_017027610.1 | XP_016883099.1 | 6 | 1535 | |||
GenBank mRNA | AK054985.1 | - | 1828 | |||
AK307459.1 | - | 961 | ||||
BC010733.1 | - | 5 | 543 | |||
BC017245.2 | - | 8 | 927 | 4890786 | ||
CR749213.1 | - | 2 | 4443 | |||
D64154.1 | - | 8 | 922 |
