siRNA Details
| Gene Symbol: BTN3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145008.1 | NP_001138480.1 | 2 | 399 | ||
| NM_001145009.1 | NP_001138481.1 | 2 | 399 | |||
| NM_007048.5 | NP_008979.3 | 2 | 399 | |||
| NM_194441.2 | NP_919423.1 | 2 | 229 | |||
| XM_005248833.2 | XP_005248890.1 | 2 | 367 | |||
| XM_005248834.3 | XP_005248891.1 | 2 | 394 | |||
| XM_006714983.2 | XP_006715046.1 | 2 | 386 | |||
| GenBank mRNA | AK223474.1 | - | 1 | 180 | ||
| AK290193.1 | - | 2 | 396 | |||
| AK291162.1 | - | 2 | 229 | |||
| AK299327.1 | - | 2 | 341 | |||
| BC118586.1 | - | 2 | 360 | 40085016 | ||
| BC121800.1 | - | 2 | 360 | 40085015 | ||
| BM147268.1 | - | 2 | 385 | |||
| BU178206.1 | - | 2 | 327 | |||
| DC417480.1 | - | 2 | 400 | |||
| T86577.1 | - | 2 | 122 | |||
| U90552.2 | - | 2 | 329 | |||
| Y07827.1 | - | 2 | 121 | |||

BTN3A1
Genomic Map