siRNA Details
| Gene Symbol: TREX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080701.3 | NP_542432.2 | 2 | 628 | ||
| GenBank mRNA | AF151107.1 | - | 1 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TREX2| Gene Symbol: TREX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080701.3 | NP_542432.2 | 2 | 628 | ||
| GenBank mRNA | AF151107.1 | - | 1 | 84 | ||