siRNA Details
| Gene Symbol: TUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007275.2 | NP_009206.1 | 3 | 448 | ||
| GenBank mRNA | AF055479.1 | - | 3 | 456 | ||
| AK124058.1 | - | 4 | 354 | |||
| AK311998.1 | - | 3 | 438 | |||
| BC023976.2 | - | 3 | 400 | 3504783 | ||
| CA392689.1 | - | 3 | 448 | |||

TUSC2
Genomic Map