siRNA Details
| Gene Symbol: FRYL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015030.1 | NP_055845.1 | 9 | 1118 | ||
| XM_005248090.4 | XP_005248147.1 | 9 | 1131 | |||
| XM_011513680.2 | XP_011511982.1 | 6 | 593 | |||
| XM_011513681.2 | XP_011511983.1 | 6 | 593 | |||
| XM_011513682.2 | XP_011511984.1 | 6 | 593 | |||
| XM_011513683.2 | XP_011511985.1 | 8 | 967 | |||
| XM_011513685.2 | XP_011511987.1 | 6 | 593 | |||
| XM_017008046.1 | XP_016863535.1 | 10 | 1443 | |||

FRYL
Genomic Map