siRNA Details
| Gene Symbol: EFR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323553.1 | NP_001310482.1 | 14 | 1811 | ||
| NM_001323554.1 | NP_001310483.1 | 14 | 2750 | |||
| NM_001323555.1 | NP_001310484.1 | 14 | 2819 | |||
| NM_001323556.1 | NP_001310485.1 | 14 | 1592 | |||
| NM_001323557.1 | NP_001310486.1 | 14 | 1661 | |||
| NM_001323558.1 | NP_001310487.1 | 14 | 1742 | |||
| NM_015137.5 | NP_055952.2 | 14 | 1742 | |||
| NR_136615.1 | 14 | 1811 | ||||
| NR_136616.1 | 14 | 1742 | ||||
| GenBank mRNA | AK294485.1 | - | 8 | 881 | ||
| AK302799.1 | - | 5 | 422 | |||
| BC071611.1 | - | 14 | 1719 | |||
| BC108667.1 | - | 14 | 1710 | |||
| BC131486.1 | - | 10 | 1077 | |||
| BC152442.1 | - | 14 | 1710 | 8862567 | ||
| BX648595.1 | - | 2 | 113 | |||
| CR749243.1 | - | 14 | 1552 | |||
| D63477.1 | - | 14 | 1710 | |||

EFR3A
Genomic Map