siRNA Details
| Gene Symbol: NBEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015175.2 | NP_055990.1 | 43 | 7069 | ||
| XM_006713072.3 | XP_006713135.1 | 42 | 6985 | |||
| XM_011533533.2 | XP_011531835.1 | 43 | 7066 | |||
| XM_017006010.1 | XP_016861499.1 | 43 | 7066 | |||
| XM_017006011.1 | XP_016861500.1 | 43 | 6922 | |||
| XM_017006012.1 | XP_016861501.1 | 42 | 6985 | |||
| XM_017006013.1 | XP_016861502.1 | 43 | 7066 | |||
| XM_017006014.1 | XP_016861503.1 | 42 | 6841 | |||
| XM_017006015.1 | XP_016861504.1 | 41 | 6697 | |||
| XM_017006016.1 | XP_016861505.1 | 38 | 7643 | |||
| XM_017006017.1 | XP_016861506.1 | 20 | 3485 | |||
| GenBank mRNA | AB011112.3 | - | 29 | 4753 | ||
| AK074036.1 | - | 1 | 903 | |||
| AK131104.1 | - | 43 | 6909 | |||
| AK293387.1 | - | 4 | 440 | |||
| AK301269.1 | - | 8 | 1132 | |||
| AY358455.1 | - | 13 | 2019 | |||
| BC029944.1 | - | 5 | 490 | |||
| BC060874.1 | - | 11 | 1547 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_183276, BC172118, BC157956, BC052551, AK129158.

NBEAL2
Genomic Map