siRNA Details
| Gene Symbol: NBEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015175.2 | NP_055990.1 | 52 | 8189 | ||
| XM_006713072.3 | XP_006713135.1 | 51 | 8105 | |||
| XM_011533533.2 | XP_011531835.1 | 51 | 8096 | |||
| XM_017006010.1 | XP_016861499.1 | 52 | 8186 | |||
| XM_017006011.1 | XP_016861500.1 | 52 | 8042 | |||
| XM_017006012.1 | XP_016861501.1 | 51 | 8105 | |||
| XM_017006013.1 | XP_016861502.1 | 51 | 8096 | |||
| XM_017006014.1 | XP_016861503.1 | 51 | 7961 | |||
| XM_017006015.1 | XP_016861504.1 | 50 | 7817 | |||
| XM_017006016.1 | XP_016861505.1 | 47 | 8763 | |||
| XM_017006017.1 | XP_016861506.1 | 29 | 4605 | |||
| GenBank mRNA | AB011112.3 | - | 38 | 5873 | ||
| AK074036.1 | - | 1 | 3520 | |||
| AK131104.1 | - | 52 | 8029 | |||
| AK293387.1 | - | 13 | 1560 | |||
| AK301269.1 | - | 17 | 2252 | |||
| AY358455.1 | - | 21 | 3049 | |||
| BC029944.1 | - | 14 | 1610 | |||
| BC060874.1 | - | 20 | 2667 | |||

NBEAL2
Genomic Map