siRNA Details
Gene Symbol: KIAA1107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000800.1 | XP_016856289.1 | 7 | 1666 | ||
GenBank mRNA | AB029030.1 | - | 6 | 1437 | ||
AK302496.1 | - | 6 | 1542 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA1107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000800.1 | XP_016856289.1 | 7 | 1666 | ||
GenBank mRNA | AB029030.1 | - | 6 | 1437 | ||
AK302496.1 | - | 6 | 1542 |