siRNA Details
| Gene Symbol: FKBP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015258.1 | NP_056073.1 | 12 | 1266 | ||
| XM_006717018.2 | XP_006717081.1 | 12 | 1183 | |||
| XM_006717019.1 | XP_006717082.1 | 11 | 1114 | |||
| XM_017014552.1 | XP_016870041.1 | 11 | 1114 | |||
| GenBank mRNA | AB014574.1 | - | 12 | 1193 | ||
| AK303074.1 | - | 11 | 1114 | |||
| AK307498.1 | - | 11 | 1095 | |||
| AK307507.1 | - | 16 | 1700 | |||
| BC009609.1 | - | 12 | 1223 | |||
| DA238568.1 | - | 3 | 246 | |||

FKBP15
Genomic Map