siRNA Details
| Gene Symbol: KIAA0930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009880.1 | NP_001009880.1 | 8 | 962 | ||
| NM_015264.1 | NP_056079.1 | 8 | 1034 | |||
| XM_011530017.2 | XP_011528319.1 | 8 | 997 | |||
| XM_017028672.1 | XP_016884161.1 | 8 | 1006 | |||
| GenBank mRNA | AB023147.1 | - | 7 | 730 | ||
| AK025608.1 | - | 6 | 551 | |||
| AK094937.1 | - | 9 | 1045 | |||
| AK124648.1 | - | 8 | 1042 | |||
| AK297058.1 | - | 8 | 1014 | |||
| AK309015.1 | - | 7 | 1086 | |||
| AK315883.1 | - | 8 | 946 | |||
| BC016882.1 | - | 3 | 292 | |||
| BC038414.1 | - | 7 | 1109 | |||
| BC053595.1 | - | 8 | 956 | 5730020 | ||
| CR456402.1 | - | 8 | 899 | |||
| KC154032.1 | - | 8 | 962 | |||

KIAA0930
Genomic Map