siRNA Details
| Gene Symbol: SMCHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015295.2 | NP_056110.2 | 25 | 3300 | ||
| XM_011525642.1 | XP_011523944.1 | 25 | 3300 | |||
| XM_011525643.2 | XP_011523945.1 | 25 | 3300 | |||
| XM_011525644.2 | XP_011523946.1 | 24 | 3208 | |||
| XM_017025684.1 | XP_016881173.1 | 26 | 3478 | |||
| GenBank mRNA | AK126324.1 | - | 10 | 931 | ||
| BU509173.1 | - | 6 | 566 | |||
| CR627458.1 | - | 14 | 1524 | |||
| DA283721.1 | - | 3 | 252 | |||
| DA675822.1 | - | 2 | 155 | |||

SMCHD1
Genomic Map