siRNA Details
| Gene Symbol: SMCHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015295.2 | NP_056110.2 | 4 | 646 | ||
| XM_011525642.1 | XP_011523944.1 | 4 | 646 | |||
| XM_011525643.2 | XP_011523945.1 | 4 | 646 | |||
| XM_011525644.2 | XP_011523946.1 | 3 | 554 | |||
| XM_017025684.1 | XP_016881173.1 | 5 | 824 | |||
| GenBank mRNA | BC006008.1 | - | 4 | 646 | ||

SMCHD1
Genomic Map