siRNA Details
| Gene Symbol: HAUS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015302.1 | NP_056117.1 | 12 | 990 | ||
| XM_011526684.2 | XP_011524986.1 | 11 | 877 | |||
| GenBank mRNA | AB020648.1 | - | 12 | 963 | ||
| BC013947.2 | - | 12 | 962 | |||
| BC064390.1 | - | 11 | 926 | |||
| BC157882.1 | - | 12 | 981 | 8862704 | ||
| BC171905.1 | - | 12 | 982 | |||

HAUS5
Genomic Map