siRNA Details
| Gene Symbol: NUP188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015354.2 | NP_056169.1 | 32 | 3507 | ||
| GenBank mRNA | BC005407.1 | - | 13 | 1448 | ||
| BC040352.1 | - | 19 | 2114 | |||
| BC111045.1 | - | 10 | 1156 | |||
| BX537774.1 | - | 29 | 3151 | |||
| D79991.1 | - | 32 | 3469 | |||

NUP188
Genomic Map