siRNA Details
| Gene Symbol: CHST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024533.4 | NP_078809.2 | 3 | 1518 | ||
| GenBank mRNA | AF246718.1 | - | 3 | 1519 | ||
| BC147002.1 | - | 1 | 343 | 9056930 | ||
| BC147007.1 | - | 1 | 343 | 9056935 | ||
| BX282540.1 | - | 4 | 357 | |||

CHST5
Genomic Map