siRNA Details
Gene Symbol: FAM215A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026770.1 | 1 | 197 | |||
GenBank mRNA | AF143236.1 | - | 1 | 209 | ||
BC069097.1 | - | 1 | 197 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM215A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026770.1 | 1 | 197 | |||
GenBank mRNA | AF143236.1 | - | 1 | 209 | ||
BC069097.1 | - | 1 | 197 |