siRNA Details
| Gene Symbol: PYY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003064.2 | 2 | 484 | |||
| GenBank mRNA | AF222904.1 | - | 2 | 233 | ||
| BC093811.1 | - | 1 | 166 | |||
| BC093813.1 | - | 1 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PYY2| Gene Symbol: PYY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003064.2 | 2 | 484 | |||
| GenBank mRNA | AF222904.1 | - | 2 | 233 | ||
| BC093811.1 | - | 1 | 166 | |||
| BC093813.1 | - | 1 | 166 | |||