siRNA Details
| Gene Symbol: PYY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003064.2 | 2 | 891 | |||
| GenBank mRNA | AF222904.1 | - | 2 | 644 | ||
| Gene Symbol: LOC105371930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011525587.1 | XP_011523889.1 | 2 | 520 | ||

PYY2
Genomic Map