siRNA Details
| Gene Symbol: KIAA1549L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012194.2 | NP_036326.2 | 4 | 2934 | ||
| XM_005252847.3 | XP_005252904.2 | 6 | 3028 | |||
| XM_005252848.3 | XP_005252905.1 | 4 | 2952 | |||
| XM_011519969.2 | XP_011518271.1 | 4 | 2952 | |||
| XM_011519970.2 | XP_011518272.1 | 5 | 1594 | |||
| XM_017017484.1 | XP_016872973.1 | 4 | 2952 | |||
| XM_017017485.1 | XP_016872974.1 | 4 | 2952 | |||
| XM_017017486.1 | XP_016872975.1 | 5 | 1594 | |||
| GenBank mRNA | AK124395.1 | - | 4 | 2952 | ||
| U10991.1 | - | 4 | 2341 | |||

KIAA1549L
Genomic Map