siRNA Details
| Gene Symbol: GSTTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003081.1 | 4 | 567 | |||
| GenBank mRNA | AL050257.1 | - | 4 | 567 | ||
| Gene Symbol: LOC100652871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011530666.1 | XP_011528968.1 | 4 | 626 | ||

GSTTP1
Genomic Map