siRNA Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002880.1 | NP_001002880.2 | 4 | 424 | ||
| NM_015373.3 | NP_056188.1 | 3 | 284 | |||
| GenBank mRNA | AB111855.1 | - | 2 | 162 | ||
| AF331041.1 | - | 3 | 220 | |||
| AK307265.1 | - | 3 | 299 | |||
| AK311928.1 | - | 3 | 297 | |||
| AL050345.1 | - | 3 | 263 | |||
| AL136686.1 | - | 3 | 267 | |||
| BC008839.2 | - | 2 | 1564 | |||
| BC016139.1 | - | 3 | 262 | 3922707 | ||
| CD702742.1 | - | 3 | 322 | |||
| CR456410.1 | - | 2 | 176 | |||

CBY1
Genomic Map