siRNA Details
| Gene Symbol: ZNF324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014347.2 | NP_055162.1 | 2, 3 | 277 | ||
| XM_005258713.4 | XP_005258770.1 | 2, 3 | 1072 | |||
| GenBank mRNA | AF060503.1 | - | 2, 3 | 226 | ||
| AK092341.1 | - | 2, 3 | 628 | |||
| BC007717.2 | - | 2, 3 | 230 | 4303415 | ||

ZNF324
Genomic Map