siRNA Details
| Gene Symbol: TPSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012467.3 | NP_036599.3 | 4 | 454 | ||
| XM_011522446.2 | XP_011520748.1 | 4 | 1193 | |||
| XM_011522447.2 | XP_011520749.1 | 4 | 2067 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPSG1| Gene Symbol: TPSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012467.3 | NP_036599.3 | 4 | 454 | ||
| XM_011522446.2 | XP_011520748.1 | 4 | 1193 | |||
| XM_011522447.2 | XP_011520749.1 | 4 | 2067 | |||