siRNA Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308053.1 | NP_001294982.1 | 9 | 814 | ||
| NM_014353.4 | NP_055168.2 | 8 | 775 | |||
| XM_011522448.1 | XP_011520750.1 | 8 | 833 | |||
| XM_011522450.2 | XP_011520752.1 | 7 | 1592 | |||
| GenBank mRNA | AB027137.1 | - | 7 | 446 | ||
| AF498952.1 | - | 7 | 443 | |||
| AK308560.1 | - | 8 | 1074 | |||
| AK314110.1 | - | 9 | 814 | |||
| AY646153.1 | - | 8 | 792 | |||
| BC066913.1 | - | 8 | 781 | 5262795 | ||
| CB242472.1 | - | 2 | 604 | |||

RAB26
Genomic Map