siRNA Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308053.1 | NP_001294982.1 | 10 | 893 | ||
| NM_014353.4 | NP_055168.2 | 9 | 854 | |||
| XM_011522448.1 | XP_011520750.1 | 9 | 912 | |||
| XM_011522450.2 | XP_011520752.1 | 8 | 1671 | |||
| GenBank mRNA | AB027137.1 | - | 8 | 525 | ||
| AF498952.1 | - | 8 | 522 | |||
| AK308560.1 | - | 9 | 1153 | |||
| AK314110.1 | - | 10 | 893 | |||
| AY646153.1 | - | 9 | 871 | |||
| BC066913.1 | - | 9 | 860 | 5262795 | ||

RAB26
Genomic Map