siRNA Details
Gene Symbol: AHCTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323342.1 | NP_001310271.1 | 2, 3 | 520 | ||
NM_001323343.1 | NP_001310272.1 | 2, 3 | 520 | |||
NM_015446.4 | NP_056261.4 | 2, 3 | 236 | |||
NR_136586.1 | 2, 3 | 520 | ||||
XM_006711758.1 | XP_006711821.1 | 2, 3 | 349 | |||
XM_011544156.2 | XP_011542458.2 | 2, 3 | 215 | |||
XM_011544157.2 | XP_011542459.2 | 2, 3 | 215 | |||
GenBank mRNA | AY157619.1 | - | 2, 3 | 354 |
