siRNA Details
| Gene Symbol: C20orf194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009984.2 | NP_001009984.1 | 25, 26 | 2348 | ||
| XM_005260684.3 | XP_005260741.1 | 25, 26 | 2342 | |||
| XM_005260687.4 | XP_005260744.1 | 6, 7 | 1184 | |||
| XM_011529208.2 | XP_011527510.1 | 6, 7 | 685 | |||
| XM_017027783.1 | XP_016883272.1 | 22, 23 | 1969 | |||
| GenBank mRNA | AK226100.1 | - | 20, 21 | 1772 | ||
| AL110249.1 | - | 16, 17 | 1313 | |||
| BC122529.1 | - | 16, 17 | 1566 | 40012183 | ||

C20orf194
Genomic Map