siRNA Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303450.1 | NP_001290379.1 | 5 | 981 | ||
| NM_021646.3 | NP_067678.1 | 5 | 981 | |||
| XM_005255243.3 | XP_005255300.1 | 4 | 947 | |||
| XM_011522453.2 | XP_011520755.1 | 5 | 982 | |||
| XM_011522454.2 | XP_011520756.1 | 4 | 736 | |||
| XM_017023121.1 | XP_016878610.1 | 4 | 474 | |||
| GenBank mRNA | AB011129.1 | - | 4 | 721 | ||
| AK291792.1 | - | 5 | 925 | |||
| AK297993.1 | - | 5 | 980 | |||
| BC121013.1 | - | 4 | 707 | 40117042 | ||
| BC121014.1 | - | 4 | 664 | 40117044 | ||

ZNF500
Genomic Map