siRNA Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303450.1 | NP_001290379.1 | 5, 6 | 1055 | ||
| NM_021646.3 | NP_067678.1 | 5, 6 | 1055 | |||
| XM_005255243.3 | XP_005255300.1 | 4, 5 | 1021 | |||
| XM_011522453.2 | XP_011520755.1 | 5, 6 | 1056 | |||
| XM_011522454.2 | XP_011520756.1 | 4, 5 | 810 | |||
| XM_017023121.1 | XP_016878610.1 | 4, 5 | 548 | |||
| GenBank mRNA | AB011129.1 | - | 4, 5 | 795 | ||
| AK291792.1 | - | 5, 6 | 999 | |||
| AK297993.1 | - | 5, 6 | 1054 | |||
| BC121013.1 | - | 4, 5 | 781 | 40117042 | ||
| BC121014.1 | - | 4, 5 | 738 | 40117044 | ||

ZNF500
Genomic Map