siRNA Details
| Gene Symbol: SLITRK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015567.1 | NP_056382.1 | 2 | 481 | ||
| XM_005254038.4 | XP_005254095.1 | 2 | 1152 | |||
| XM_005254039.2 | XP_005254096.1 | 2 | 821 | |||
| GenBank mRNA | AB020725.1 | - | 2 | 287 | ||
| AK024251.1 | - | 2 | 481 | |||
| BC098106.1 | - | 2 | 389 | 40012375 | ||
| BC098287.1 | - | 2 | 389 | 40012379 | ||
| BC103509.1 | - | 2 | 389 | 40012381 | ||

SLITRK5
Genomic Map