siRNA Details
Gene Symbol: FAM198A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129908.2 | NP_001123380.2 | 2 | 594 | ||
GenBank mRNA | AK091001.1 | - | 2 | 524 | ||
AK311059.1 | - | 2 | 267 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM198A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129908.2 | NP_001123380.2 | 2 | 594 | ||
GenBank mRNA | AK091001.1 | - | 2 | 524 | ||
AK311059.1 | - | 2 | 267 |