siRNA Details
| Gene Symbol: GAS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038733.1 | - | 2 | 52 | ||
| BC070486.1 | - | 1 | 17 | |||
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DA173140.1 | - | 2 | 52 | ||

GAS5
Genomic Map