siRNA Details
| Gene Symbol: DENND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318052.1 | NP_001304981.1 | 11 | 2405 | ||
| NM_001318053.1 | NP_001304982.1 | 12 | 2315 | |||
| NM_015689.4 | NP_056504.3 | 12 | 2507 | |||
| NR_134477.1 | 10 | 2132 | ||||
| XM_005249977.3 | XP_005250034.1 | 12 | 3038 | |||
| XM_005249979.4 | XP_005250036.1 | 12 | 2291 | |||
| XM_011516052.2 | XP_011514354.1 | 12 | 2385 | |||
| XM_011516053.2 | XP_011514355.1 | 11 | 2489 | |||
| XM_011516054.2 | XP_011514356.1 | 12 | 2290 | |||
| XM_017011989.1 | XP_016867478.1 | 11 | 2491 | |||
| GenBank mRNA | AB033103.2 | - | 11 | 2405 | ||
| AK290056.1 | - | 12 | 2276 | |||
| BC049193.1 | - | 8 | 875 | |||
| BC115004.1 | - | 12 | 2315 | 7470138 | ||
| BC132875.1 | - | 10 | 2102 | 40146897 | ||

DENND2A
Genomic Map