siRNA Details
| Gene Symbol: C5AR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271749.1 | NP_001258678.1 | 2 | 605 | ||
| NM_001271750.1 | NP_001258679.1 | 2 | 528 | |||
| NM_018485.2 | NP_060955.1 | 2 | 567 | |||
| XM_011526736.2 | XP_011525038.1 | 3 | 1242 | |||
| GenBank mRNA | AB038237.1 | - | 2 | 458 | ||
| AK313994.1 | - | 2 | 605 | |||
| BC067478.1 | - | 1 | 406 | 6971812 | ||
| BC067479.1 | - | 1 | 406 | 6971813 | ||
| BC067480.1 | - | 1 | 406 | 6971814 | ||
| BP370117.1 | - | 2 | 528 | |||
More Information
Set Membership: |
|

C5AR2
Genomic Map