siRNA Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014170.2 | NP_054889.2 | 1 | 245 | ||
| NM_138485.1 | NP_612494.1 | 1 | 245 | |||
| GenBank mRNA | AF161484.1 | - | 1 | 215 | ||
| AK289611.1 | - | 1 | 207 | |||
| AK292651.1 | - | 1 | 264 | |||
| BC010137.2 | - | 1 | 211 | |||
| BC037163.1 | - | 1 | 248 | 4620206 | ||
| BG655089.1 | - | 1 | 189 | |||
| BI830308.1 | - | 1 | 245 | |||
| BX354218.2 | - | 1 | 213 | |||
| CK818022.1 | - | 1 | 176 | |||

GTPBP8
Genomic Map