siRNA Details
| Gene Symbol: UQCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003684.1 | NP_001003684.1 | 2 | 724 | ||
| NM_013387.3 | NP_037519.2 | 2 | 665 | |||
| GenBank mRNA | BI754905.1 | - | 2 | 627 | ||
| CA311834.1 | - | 2 | 266 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UQCR10| Gene Symbol: UQCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003684.1 | NP_001003684.1 | 2 | 724 | ||
| NM_013387.3 | NP_037519.2 | 2 | 665 | |||
| GenBank mRNA | BI754905.1 | - | 2 | 627 | ||
| CA311834.1 | - | 2 | 266 | |||